Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AH014147.3 | - | 3 | 1109 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |