Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002965.2 | NP_001002965.2 | 1083 | 113 | ||
NM_001293224.1 | NP_001280153.1 | 1 | 963 | 113 | ||
GenBank mRNA | AF544242.1 | - | 1 | 1087 | 113 | |
DN875727.1 | - | 1 | 325 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |