Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109813.1 | NP_001103283.1 | 3 - 4 | 1345 | 87 | |
XM_005622263.3 | XP_005622320.1 | 2 - 3 | 1248 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |