Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109813.1 | NP_001103283.1 | 5 - 6 | 1809 | 79 | |
XM_005622263.3 | XP_005622320.1 | 4 - 5 | 1712 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |