Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159762.2 | NP_001153234.1 | 1 - 2 | 173 | 82 | |
GenBank mRNA | CX993709.1 | - | 3 - 4 | 298 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |