Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185003.1 | NP_001171932.1 | 1 - 2 | 134 | 64 | |
XM_022427141.1 | XP_022282849.1 | 1 - 2 | 161 | 64 | ||
GenBank mRNA | FN825779.1 | - | 1 - 2 | 134 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |