Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197035.2 | NP_001183964.2 | 1 - 2 | 975 | 61 | |
GenBank mRNA | X14050.1 | - | 664 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris Probe spans exons |