Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205190.2 | NP_001192119.1 | 6 - 7 | 898 | 67 | |
XM_005633260.3 | XP_005633317.1 | 7 - 8 | 1049 | 67 | ||
XM_014122406.2 | XP_013977881.1 | 7 - 8 | 1041 | 67 | ||
XM_022407407.1 | XP_022263115.1 | 8 - 9 | 1206 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |