Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252348.1 | NP_001239277.1 | 4 - 5 | 351 | 108 | |
XM_005633838.3 | XP_005633895.1 | 7 - 8 | 1295 | 108 | ||
XM_014106255.2 | XP_013961730.1 | 7 - 8 | 1308 | 108 | ||
XM_022408005.1 | XP_022263713.1 | 7 - 8 | 1295 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |