Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254715.1 | NP_001241644.1 | 3 - 4 | 477 | 58 |
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417343.1 | XP_022273051.1 | 5 - 6 | 920 | 58 | |
XM_022417344.1 | XP_022273052.1 | 5 - 6 | 800 | 58 | ||
XM_022417347.1 | XP_022273055.1 | 4 - 5 | 860 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |