Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246775.1 | NP_001233704.1 | 3 - 4 | 379 | 70 | |
GenBank mRNA | AF128242.1 | - | 3 - 4 | 399 | 70 | |
JI871500.1 | - | 3 - 4 | 379 | 70 | ||
JP047112.1 | - | 3 - 4 | 379 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |