Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246800.1 | NP_001233729.1 | 3 - 4 | 788 | 80 | |
GenBank mRNA | FJ476073.1 | - | 3 - 4 | 788 | 80 | |
JI895720.1 | - | 3 - 4 | 549 | 80 | ||
JP057040.1 | - | 3 - 4 | 549 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |