Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246805.1 | NP_001233734.1 | 1 - 2 | 814 | 108 | |
GenBank mRNA | AB022191.1 | - | 1 - 2 | 814 | 108 | |
JI886486.1 | - | 1 - 2 | 753 | 108 | ||
JP056670.1 | - | 1 - 2 | 751 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons |