Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285924.1 | NP_001272853.1 | 2 - 3 | 162 | 75 | |
GenBank mRNA | JI871282.1 | - | 1 - 2 | 286 | 75 | |
JP044871.1 | - | 2 - 3 | 162 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |