Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243990.1 | NP_001230919.1 | 3 - 4 | 447 | 102 | |
GenBank mRNA | AB042238.1 | - | 3 - 4 | 447 | 102 | |
JI877077.1 | - | 3 - 4 | 382 | 102 | ||
JP061058.1 | - | 3 - 4 | 382 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |