Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244056.1 | NP_001230985.1 | 3 - 4 | 457 | 68 | |
GenBank mRNA | DQ191064.1 | - | 3 - 4 | 457 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |