Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246714.1 | NP_001233643.1 | 1 - 2 | 39 | 68 | |
GenBank mRNA | AB271736.1 | - | 1 - 2 | 39 | 68 | |
JI875398.1 | - | 1 - 2 | 125 | 68 | ||
JP037636.1 | - | 1 - 2 | 125 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |