Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503715.2 | XP_003503763.1 | 1 - 2 | 63 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |