Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649008.1 | XP_007647198.1 | 3 - 4 | 511 | 66 | |
GenBank mRNA | JI869845.1 | - | 3 - 4 | 295 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |