Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514176.2 | XP_003514224.1 | 4 - 5 | 392 | 60 | |
GenBank mRNA | JI885411.1 | - | 1 - 2 | 78 | 60 | |
JP038857.1 | - | 1 - 2 | 78 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |