Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503147.2 | XP_003503195.2 | 9 - 10 | 985 | 63 | |
GenBank mRNA | JI879019.1 | - | 2 - 3 | 249 | 63 | |
JP056190.1 | - | 2 - 3 | 249 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |