Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506723.2 | XP_003506771.2 | 1 - 2 | 230 | 88 | |
GenBank mRNA | JP049778.1 | - | 1 - 2 | 103 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |