Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501843.2 | XP_003501891.1 | 1 - 2 | 130 | 74 | |
GenBank mRNA | JI871166.1 | - | 1 - 2 | 58 | 74 |