Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514588.2 | XP_003514636.1 | 3 - 4 | 381 | 97 | |
GenBank mRNA | JI870243.1 | - | 3 - 4 | 358 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |