Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508684.2 | XP_003508732.1 | 2 - 3 | 1018 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |