Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513798.2 | XP_003513846.1 | 18 - 19 | 2302 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |