Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643701.1 | XP_007641891.1 | 1 - 2 | 170 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |