Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl24d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495020.2 | XP_003495068.1 | 2 - 3 | 267 | 80 | |
GenBank mRNA | JI886744.1 | - | 2 - 3 | 259 | 80 | |
JP039861.1 | - | 2 - 3 | 259 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |