Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499172.2 | XP_003499220.1 | 2 - 3 | 306 | 105 | |
XM_007638866.1 | XP_007637056.1 | 2 - 3 | 306 | 105 | ||
XM_007638873.1 | XP_007637063.1 | 2 - 3 | 306 | 105 | ||
XM_007638881.1 | XP_007637071.1 | 2 - 3 | 306 | 105 | ||
XM_007638890.1 | XP_007637080.1 | 2 - 3 | 306 | 105 | ||
GenBank mRNA | JI871025.1 | - | 2 - 3 | 185 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |