Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512751.2 | XP_003512799.1 | 2 - 3 | 307 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |