Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640146.1 | XP_007638336.1 | 4 - 5 | 491 | 58 | |
GenBank mRNA | JI889538.1 | - | 4 - 5 | 491 | 58 | |
JP065499.1 | - | 4 - 5 | 491 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |