Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100754573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654419.1 | XP_007652609.1 | 5 - 6 | 2091 | 92 | |
GenBank mRNA | JI884333.1 | - | 1 - 2 | 496 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |