Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506360.2 | XP_003506408.1 | 1 - 2 | 132 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |