Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007622134.1 | XP_007620324.1 | 16 - 17 | 1820 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |