Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100774181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502097.2 | XP_003502145.1 | 6 - 7 | 868 | 82 | |
GenBank mRNA | JI876754.1 | - | 6 - 7 | 869 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |