Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501581.2 | XP_003501629.1 | 9 - 10 | 2676 | 87 | |
XM_007642232.1 | XP_007640422.1 | 8 - 9 | 1550 | 87 | ||
XM_007642233.1 | XP_007640423.1 | 8 - 9 | 1313 | 87 | ||
XM_007642234.1 | XP_007640424.1 | 7 - 8 | 1197 | 87 | ||
XM_007642235.1 | XP_007640425.1 | 8 - 9 | 1269 | 87 | ||
XM_007642236.1 | XP_007640426.1 | 8 - 9 | 1224 | 87 | ||
GenBank mRNA | JI877155.1 | - | 2 - 3 | 343 | 87 | |
JP047302.1 | - | 2 - 3 | 343 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |