Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505836.2 | XP_003505884.1 | 1 - 2 | 134 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |