Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JI879061.1 | - | 1 - 2 | 79 | ||
JP042737.1 | - | 1 - 2 | 124 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |