Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633880.1 | XP_007632070.1 | 16 - 17 | 3268 | 98 | |
XM_007633884.1 | XP_007632074.1 | 16 - 17 | 2958 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |