Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609741.1 | XP_007607931.1 | 5 - 6 | 788 | 60 | |
GenBank mRNA | JP042244.1 | - | 5 - 6 | 794 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |