Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497471.2 | XP_003497519.1 | 1 - 2 | 276 | 94 | |
GenBank mRNA | JI895394.1 | - | 1 - 2 | 208 | 94 | |
JP053247.1 | - | 1 - 2 | 209 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |