Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506873.2 | XP_003506921.1 | 2 - 3 | 185 | 70 | |
GenBank mRNA | JI868678.1 | - | 2 - 3 | 110 | 70 | |
JP042089.1 | - | 2 - 3 | 110 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |