Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510893.1 | XP_003510941.1 | 3 - 4 | 131 | 96 | |
XM_007651495.1 | XP_007649685.1 | 3 - 4 | 134 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |