Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499533.2 | XP_003499581.1 | 3 - 4 | 787 | 77 | |
XM_007640297.1 | XP_007638487.1 | 3 - 4 | 538 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |