Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503484.2 | XP_003503532.1 | 4 - 5 | 816 | 58 | |
GenBank mRNA | JI879404.1 | - | 3 - 4 | 643 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |