Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647241.1 | XP_007645431.1 | 3 - 4 | 489 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |