Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653957.1 | XP_007652147.1 | 10 - 11 | 1377 | 90 | |
XM_007653960.1 | XP_007652150.1 | 10 - 11 | 1129 | 90 | ||
XM_007653965.1 | XP_007652155.1 | 10 - 11 | 1099 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Protein Kinase Detect Genomic DNA Probe spans exons |