Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644295.1 | XP_007642485.1 | 6 - 7 | 2131 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |