Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499418.2 | XP_003499466.2 | 5 - 6 | 862 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |