Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508005.1 | XP_003508053.1 | 1 | 1020 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Both primers and probe map within a single exon |